MM-102 ≥99%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(Bis(4-fluorophenyl)methyl)-1-(2-(2-ethyl-2-isobutyramidobutanamido)-5-guanidinopentanamido)cyclopentanecarboxamide, 1-[[(2S)-5-[(aminoiminomethyl)amino]-2-[[2-ethyl-2-[(2-methyl-1-oxopropyl)amino]-1-oxobutyl]amino]-1-oxopentyl]amino]-N-[bis(4-fluorophenyl)methyl]-cyclopentanecarboxamide

A622764-1MG A622764-5MG
77617-622EA 48.41 USD
77617-622 77747-285
MM-102 ≥99%
MM-102

Precaución: for Research Use Only

Formula: C₃₅H₄₉F₂N₇O₄
MW: 669.80 g/mol
MDL Number: MFCD28015099
CAS Number: 1417329-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR