Difluoro(4-fluorophenyl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluoro-2-(4-fluorophenyl)acetic Acid

A622585-250MG A622585-100MG A622585-1G A622585-5G
77686-563EA 29 USD
77686-563 77901-747 77900-806 77901-889
Difluoro(4-fluorophenyl)acetic acid ≥98%
Difluoro(4-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD11167795
CAS Number: 94010-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR