1,4-Anthraquinone ≥95%

Supplier: Ambeed
Danger

Synonyms: Anthracene-1,4-dione

A621981-25G A621981-5G
77669-680EA 182 USD
77669-680 77669-682
1,4-Anthraquinone ≥95%
1,4-Anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₈O₂
MW: 208.22 g/mol
MDL Number: MFCD00013724
CAS Number: 635-12-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR