Trypan blue 70% (dye content)

Supplier: Ambeed
Danger

A621935-5G A621935-25G
77613-932EA 15.45 USD
77613-932 77901-265
Trypan blue 70% (dye content)
Trypan Blue

Precaución: for Research Use Only

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number: MFCD00003969
CAS Number: 72-57-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR