Varlitinib ≥99%

Supplier: Ambeed
Danger

Synonyms: (R)-N4-(3-chloro-4-(thiazol-2-ylmethoxy)phenyl)-N6-(4-methyl-4,5-dihydrooxazol-2-yl)quinazoline-4,6-diamine

A621686-10MG A621686-2MG A621686-5MG
77686-559EA 205 USD
77686-559 77686-560 77686-561
Varlitinib ≥99%
Varlitinib

Precaución: for Research Use Only

Formula: C₂₂H₁₉ClN₆O₂S
MW: 466.94 g/mol
MDL Number: MFCD20275099
CAS Number: 845272-21-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR