1-(tert-Butyldimethylsilyl)indole-5-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1-(tert-Butyldimethylsilyl)-1H-indol-5-yl)boronic acid

A621100-250MG A621100-5G A621100-1G
77873-014EA 33 USD
77873-014 77873-015 77873-013
1-(tert-Butyldimethylsilyl)indole-5-boronic acid ≥98%
1-(tert-Butyldimethylsilyl)indole-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂Si
MDL Number: MFCD03095156
CAS Number: 913835-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR