5-Acetylsalicylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-2-hydroxybenzoic acid, 5-Acetylsalicylic Acid

A620775-1G A620775-5G
77617-616EA 21.63 USD
77617-616 77617-618
5-Acetylsalicylic acid ≥98%
5-Acetylsalicylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MDL Number: MFCD00013978
CAS Number: 13110-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR