5,6-Dichloro-1,3-dihydro-2H-benzimidazol-2-one ≥98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-1H-benzo[d]imidazol-2(3H)-one

A620269-100MG A620269-1G
77884-480EA 13 USD
77884-480 77884-481
5,6-Dichloro-1,3-dihydro-2H-benzimidazol-2-one ≥98%
5,6-Dichloro-1,3-dihydro-2H-benzimidazol-2-one

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂O
MW: 203.03 g/mol
MDL Number: MFCD12963764
CAS Number: 2033-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR