1-Boc-3-(hydroxymethyl)-3-methylazetidine ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)-3-methylazetidine, N-BOC protected, tert-Butyl 3-(hydroxymethyl)-3-methylazetidine-1-carboxylate

A620211-100MG A620211-1G A620211-250MG
77673-672EA 47 USD
77673-672 77673-674 77673-676
1-Boc-3-(hydroxymethyl)-3-methylazetidine ≥97%
1-Boc-3-(hydroxymethyl)-3-methylazetidine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD20926177
CAS Number: 1363382-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR