tert-Butyl (3-aminopropyl)(4-((tert-butoxycarbonyl)amino)butyl)carbamate ≥95%

Supplier: Ambeed
Warning

A618992-1G A618992-250MG
77900-686EA 518 USD
77900-686 77900-936
tert-Butyl (3-aminopropyl)(4-((tert-butoxycarbonyl)amino)butyl)carbamate ≥95%
tert-Butyl (3-aminopropyl)(4-((tert-butoxycarbonyl)amino)butyl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₃₅N₃O₄
MDL Number: MFCD04973457
CAS Number: 68076-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR