trans-2-Aminocyclobutanol hydrochloride ≥97%

Supplier: Ambeed
Warning

A618115-5G A618115-10G
77594-906EA 1947.5 USD
77594-906 77747-277
trans-2-Aminocyclobutanol hydrochloride ≥97%
trans-2-Aminocyclobutanol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD26127776
CAS Number: 1609406-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR