Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97%

Supplier: Ambeed
Warning

A617558-5G A617558-1G
77613-926EA 61.8 USD
77613-926 77673-664
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate ≥97%
Benzyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO₃
MDL Number: MFCD11976640
CAS Number: 77211-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR