4-((tert-Butoxycarbonyl)amino)butyl 4-methylbenzenesulfonate ≥95%

Supplier: Ambeed
Warning

Synonyms: Tos-O-C4-NH-Boc

A617319-1G A617319-250MG A617319-100MG
77613-922EA 203.94 USD
77613-922 77613-924 77686-552
4-((tert-Butoxycarbonyl)amino)butyl 4-methylbenzenesulfonate ≥95%
4-((tert-Butoxycarbonyl)amino)butyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₆H₂₅NO₅S
MW: 343.44 g/mol
MDL Number: MFCD18839142
CAS Number: 180851-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR