GDC-0349 ≥98%

Supplier: Ambeed
Warning

A617285-5MG A617285-10MG A617285-1MG
77617-602EA 60.36 USD
77617-602 77617-598 77617-600
GDC-0349 ≥98%
GDC-0349

Precaución: for Research Use Only

Formula: C₂₄H₃₂N₆O₃
MDL Number: MFCD22417087
CAS Number: 1207360-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR