Diethyl (aminomethyl)phosphonate ≥98%
Supplier: Ambeed
|
Danger
|
A616938-100MG
A616938-250MG
77884-475EA
38
USD
77884-475
77884-476
Diethyl (aminomethyl)phosphonate ≥98%
Diethyl (aminomethyl)phosphonate
Precaución: for Research Use Only
Formula:
C₅H₁₄NO₃P |
MDL Number:
MFCD01072445 CAS Number: 50917-72-1 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...