1-(4-Chlorophenyl)-3-(5-isopropyl-1-methyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)urea ≥98%

Supplier: Ambeed
Warning

Synonyms: FPR Agonist 43

A616899-10MG A616899-1MG A616899-5MG
77617-592EA 122.57 USD
77617-592 77617-594 77617-596
1-(4-Chlorophenyl)-3-(5-isopropyl-1-methyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)urea ≥98%
1-(4-Chlorophenyl)-3-(5-isopropyl-1-methyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)urea

Precaución: for Research Use Only

Formula: C₂₀H₂₁ClN₄O₂
MW: 384.86 g/mol
MDL Number: MFCD30182236
CAS Number: 903895-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR