(2-Cyclopropyl-4-methoxyphenyl)boronic acid ≥97%

Supplier: Ambeed
Warning

A616697-1G A616697-250MG
77893-240EA 634 USD
77893-240 77893-241
(2-Cyclopropyl-4-methoxyphenyl)boronic acid ≥97%
(2-Cyclopropyl-4-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MDL Number: MFCD21099644
CAS Number: 2225173-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR