Sulfo-Cyanine 3 carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-[3-[1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene]prop-1-en-1-yl]-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate, Cy3, 2-(3-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulphoindolin-2-ylidene)prop-1-en-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulphonate

A616603-1MG A616603-5MG A616603-10MG
77613-916EA 16.48 USD
77613-916 77613-918 77686-551
Sulfo-Cyanine 3 carboxylic acid ≥98%
Sulfo-Cyanine 3 carboxylic acid

Precaución: for Research Use Only

Formula: C₃₁H₃₈N₂O₈S₂
MW: 630.77 g/mol
MDL Number: MFCD28978247
CAS Number: 146368-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR