Methyl 3-bromo-5-iodobenzoate ≥98%

Supplier: Ambeed
Warning

A616566-5G A616566-1G
77573-008EA 15 USD
77573-008 77669-648
Methyl 3-bromo-5-iodobenzoate ≥98%
Methyl 3-bromo-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
MDL Number: MFCD00673016
CAS Number: 188813-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR