2-Chloro-5-ethenylpyridine ≥95% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with TBC

A616173-1G A616173-5G
77613-912EA 54.59 USD
77613-912 77891-226
2-Chloro-5-ethenylpyridine ≥95% stabilized
2-Chloro-5-ethenylpyridine

Precaución: for Research Use Only

Formula: C₇H₆ClN
MDL Number: MFCD11870540
CAS Number: 157670-28-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR