5-Hydroxy-7-methoxy-3-(4-methoxyphenyl)-4H-chromen-4-one ≥97%

Supplier: Ambeed
Warning

Synonyms: 7-O-Methylbiochanin A

A615481-250MG A615481-1G
77602-552EA 52.28 USD
77602-552 77669-642
5-Hydroxy-7-methoxy-3-(4-methoxyphenyl)-4H-chromen-4-one ≥97%
5-Hydroxy-7-methoxy-3-(4-methoxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₅
MW: 298.29 g/mol
MDL Number: MFCD00075976
CAS Number: 34086-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR