1,2-Dipalmitoyl-sn-glycerol ≥97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Hydroxypropane-1,2-diyl dipalmitate

A614374-1G A614374-5G A614374-250MG
77617-572EA 183.34 USD
77617-572 77617-574 77747-270
1,2-Dipalmitoyl-sn-glycerol ≥97%
1,2-Dipalmitoyl-sn-glycerol

Precaución: for Research Use Only

Formula: C₃₅H₆₈O₅
MW: 568.924 g/mol
MDL Number: MFCD00067471
CAS Number: 30334-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR