6,7,12b,13-Tetrahydro-4H-[1,3]dioxolo[4',5':7,8]isoquinolino[3,2-a][1,3]dioxolo[4,5-g]isoquinoline ≥98%

Supplier: Ambeed
Warning

Synonyms: (±)-Stylopine

A614191-5MG A614191-10MG A614191-1MG
77617-570EA 40.17 USD
77617-570 77686-547 77686-548
6,7,12b,13-Tetrahydro-4H-[1,3]dioxolo[4',5':7,8]isoquinolino[3,2-a][1,3]dioxolo[4,5-g]isoquinoline ≥98%
6,7,12b,13-Tetrahydro-4H-[1,3]dioxolo[4',5':7,8]isoquinolino[3,2-a][1,3]dioxolo[4,5-g]isoquinoline

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₄
MW: 323.34 g/mol
MDL Number: MFCD00075989
CAS Number: 4312-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR