5-Bromo-3-methyl-3,4-dihydroquinazolin-2(1H)-one ≥97%

Supplier: Ambeed
Warning

A614188-100MG A614188-250MG
77617-566EA 70.04 USD
77617-566 77617-568
5-Bromo-3-methyl-3,4-dihydroquinazolin-2(1H)-one ≥97%
5-Bromo-3-methyl-3,4-dihydroquinazolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MDL Number: MFCD27930245
CAS Number: 861106-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR