Piperidin-4-yl-acetic acid tert-butyl ester ≥96%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 2-(piperidin-4-yl)acetate, tert-butyl 2-(4-piperidyl)acetate

A613961-1G A613961-5G
77684-492EA 149 USD
77684-492 77684-493
Piperidin-4-yl-acetic acid tert-butyl ester ≥96%
Piperidin-4-yl-acetic acid tert-butyl ester

Precaución: For research use only.

Formula: C₁₁H₂₁NO₂
MW: 199.29 g/mol
MDL Number: MFCD06738957
CAS Number: 180182-07-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR