(4-Fluoro-2-methoxyphenyl)acetic acid ≥98%

Supplier: Ambeed
Warning

A613937-1G A613937-5G
77901-936EA 95 USD
77901-936 77901-554
(4-Fluoro-2-methoxyphenyl)acetic acid ≥98%
(4-Fluoro-2-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD06660193
CAS Number: 886498-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR