α,α-Difluoro-2-(4-methoxyphenyl)acetic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluoro-2-(4-methoxyphenyl)acetic acid

A613769-100MG A613769-1G
77891-220EA 60 USD
77891-220 77891-221
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid ≥95%
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MDL Number: MFCD11007700
CAS Number: 1027513-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR