PDGF receptor tyrosine kinase inhibitor IV ≥97%

Supplier: Ambeed
Warning

Synonyms: N-(3-Fluorophenyl)-6,7-dimethoxy-1,4-dihydroindeno[1,2-c]pyrazol-3-amine, JNJ-10198409, RWJ 540973, 3-Fluoro-N-(6,7-dimethoxy-2,4-dihydroindeno[1,2-c]pyrazol-3-yl)phenylamine, N-(3-fluorophenyl)-2,4-dihydro-6,7-dimethoxy-Indeno[1,2-c]pyrazol-3-amine

A613718-1MG A613718-5MG
77617-564EA 44.26 USD
77617-564 77747-269
PDGF receptor tyrosine kinase inhibitor IV ≥97%
PDGF receptor tyrosine kinase inhibitor IV

Precaución: for Research Use Only

Formula: C₁₈H₁₆FN₃O₂
CAS Number: 627518-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR