(R)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride ≥95%

Supplier: Ambeed
Warning

A613156-1G A613156-250MG
77893-236EA 246 USD
77893-236 77893-237
(R)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride ≥95%
(R)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂O₂
MDL Number: MFCD09878827
CAS Number: 1212327-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR