4-Bromobenzenesulfonyl fluoride ≥98%

Supplier: Ambeed
Danger

A612780-1G A612780-25G A612780-5G
77902-047EA 24 USD
77902-047 77901-549 77901-534
4-Bromobenzenesulfonyl fluoride ≥98%
4-Bromobenzenesulfonyl fluoride

Precaución: for Research Use Only

Formula: C₆H₄BrFO₂S
MW: 239.06 g/mol
MDL Number: MFCD21892326
CAS Number: 498-83-9
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR