3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 1-oxo-7-azaspiro[3.5]nonane-7-carboxylate
A612603-100MG
A612603-1G
A612603-250MG
77613-906EA
19.57
USD
77613-906
77613-908
77684-488
3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester ≥98%
3-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester
Precaución: For research use only.
Formula:
C₁₃H₂₁NO₃ MW: 239.32 g/mol |
MDL Number:
MFCD14585368 CAS Number: 849203-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...