Dirithromycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R,2R,3R,6R,7S,8S,9R,10R,12R,13S,15R,17S)-9-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-3-ethyl-2,10-dihydroxy-7-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-15-((2-methoxyethoxy)methyl)-2,6,8,10,12,17-hexamethyl-4,16-dioxa-14-azabicyclo[11.3.1]heptadecan-5-one, [9S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxyethoxy)ethylidene]oxy]erythromycin, LY-237216

A612277-5G A612277-250MG A612277-25G
77602-550EA 21.53 USD
77602-550 77627-246 77627-248
Dirithromycin ≥98%
Dirithromycin

Precaución: for Research Use Only

Formula: C₄₂H₇₈N₂O₁₄
MW: 835.09 g/mol
MDL Number: MFCD00865041
CAS Number: 62013-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR