Methyl 2-iodo-4-nitrobenzoate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A612167-1G A612167-250MG A612167-5G
77684-487EA 55 USD
77684-487 77891-212 77891-213
Methyl 2-iodo-4-nitrobenzoate ≥95%, Ambeed.Inc
Methyl 2-iodo-4-nitrobenzoate

Precaución: For research use only.

Formula: C₈H₆INO₄
MDL Number: MFCD00277928
CAS Number: 6326-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR