N3,N3,N7,N7-Tetramethyl-10H-phenothiazine-3,7-diamine dimethanesulfonate ≥98%

Supplier: Ambeed
Warning

Synonyms: Leucomethylene blue mesylate

A611795-10MG A611795-5MG
77747-259EA 29 USD
77747-259 77747-260
N3,N3,N7,N7-Tetramethyl-10H-phenothiazine-3,7-diamine dimethanesulfonate ≥98%
N3,N3,N7,N7-Tetramethyl-10H-phenothiazine-3,7-diamine dimethanesulfonate

Precaución: for Research Use Only

Formula: C₁₈H₂₇N₃O₆S₃
MDL Number: MFCD29764308
CAS Number: 1236208-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR