(±)-Blebbistatin 98
Supplier: Ambeed
|
Warning
|
Synonyms:
3a-Hydroxy-6-methyl-1-phenyl-1,2,3,3a-tetrahydro-4H-pyrrolo[2,3-b]quinolin-4-one, Blebbistatin
A611342-1MG
A611342-5MG
A611342-10MG
77617-556EA
18.54
USD
77617-556
77747-257
77747-256
(±)-Blebbistatin 98
(±)-Blebbistatin
Precaución: for Research Use Only
Formula:
C₁₈H₁₆N₂O₂ MW: 292.33 g/mol |
MDL Number:
MFCD04113775 CAS Number: 674289-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...