3-Bromo-2,4-dimethylaniline ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2,4-dimethyl-benzenamine

A611164-5G A611164-1G
77572-990EA 193.64 USD
77572-990 77684-485
3-Bromo-2,4-dimethylaniline ≥95%
3-Bromo-2,4-dimethylaniline

Precaución: For research use only.

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD20528102
CAS Number: 66314-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR