Potassium (3-(tert-butoxycarbonyl)-3-azabicyclo[3.1.0]hexan-1-yl)trifluoroborate ≥97%

Supplier: Ambeed
Warning

A611055-1G A611055-250MG A611055-100MG
77783-504EA 965 USD
77783-504 77783-505 77783-503
Potassium (3-(tert-butoxycarbonyl)-3-azabicyclo[3.1.0]hexan-1-yl)trifluoroborate ≥97%
Potassium (3-(tert-butoxycarbonyl)-3-azabicyclo[3.1.0]hexan-1-yl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₆BF₃KNO₂
MDL Number: MFCD31556357
CAS Number: 2095504-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR