N-(2-Hydroxypropyl)-2-methylprop-2-enamide ≥97%

Supplier: Ambeed
Warning

Synonyms: N-(2-Hydroxypropyl)methacrylamide

A611003-25G A611003-10G
77572-986EA 197.76 USD
77572-986 77627-244
N-(2-Hydroxypropyl)-2-methylprop-2-enamide ≥97%
N-(2-Hydroxypropyl)-2-methylprop-2-enamide

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.184 g/mol
MDL Number: MFCD00080531
CAS Number: 21442-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR