(S)-1-(2,4-Dichlorophenyl)ethanamine ≥95%

Supplier: Ambeed
Warning

A610979-250MG A610979-100MG
77878-417EA 59 USD
77878-417 77878-416
(S)-1-(2,4-Dichlorophenyl)ethanamine ≥95%
(S)-1-(2,4-Dichlorophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N
MDL Number: MFCD06761787
CAS Number: 133492-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR