EDTA tetrasodium salt tetrahydrate ≥99%

Supplier: Ambeed
Danger

Synonyms: EDTA tetrasodium tetrahydrate, Tetrasodium ethylenediaminetetraacetate tetrahydrate

A610366-500G A610366-100G
77598-604EA 6.15 USD
77598-604 77598-602
EDTA tetrasodium salt tetrahydrate ≥99%
EDTA tetrasodium salt tetrahydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₈Na₄·4H₂O
MW: 452.23 g/mol
MDL Number: MFCD00150027
CAS Number: 13235-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR