(S)-tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A610163-1G A610163-250MG
77686-540EA 246 USD
77686-540 77686-541
(S)-tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate ≥95%
(S)-tert-Butyl 3-(2-hydroxyethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.30 g/mol
MDL Number: MFCD18633590
CAS Number: 1273577-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR