4-Fluoro-2-iodo-5-methylaniline ≥97%
Supplier: Ambeed
|
Warning
|
A609757-1G
A609757-5G
77684-482EA
107
USD
77684-482
77684-483
4-Fluoro-2-iodo-5-methylaniline ≥97%
4-Fluoro-2-iodo-5-methylaniline
Precaución: For research use only.
Formula:
C₇H₇FIN MW: 251.04 g/mol |
MDL Number:
MFCD11226280 CAS Number: 85233-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...