4-Methylaminomethylphenylboronic acid ≥95%

Supplier: Ambeed
Warning

A609545-100MG A609545-250MG A609545-5G
77886-461EA 70 USD
77886-461 77886-462 77886-463
4-Methylaminomethylphenylboronic acid ≥95%
4-Methylaminomethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₂
MDL Number: MFCD03094948
CAS Number: 518336-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR