Methyl 5-(aminomethyl)picolinate dihydrochloride ≥96%

Supplier: Ambeed
Warning

A609496-1G A609496-250MG A609496-100MG A609496-5G
77572-984EA 115.36 USD
77572-984 77598-600 77617-546 77749-256
Methyl 5-(aminomethyl)picolinate dihydrochloride ≥96%
Methyl 5-(aminomethyl)picolinate dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂O₂
MW: 239.1 g/mol
MDL Number: MFCD21602618
CAS Number: 1375471-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR