5-Fluoro-1,3-dimethyl-2-nitrobenzene ≥98%

Supplier: Ambeed
Warning

A609290-10G A609290-1G A609290-5G A609290-25G
77594-866EA 213.2 USD
77594-866 77613-892 77673-652 77884-465
5-Fluoro-1,3-dimethyl-2-nitrobenzene ≥98%
5-Fluoro-1,3-dimethyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD03413407
CAS Number: 315-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR