3-Chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A608888-1G A608888-250MG
77873-001EA 64 USD
77873-001 77873-002
3-Chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride ≥98%
3-Chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃O₂S
MDL Number: MFCD13185471
CAS Number: 875167-01-4
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR