Tantalum(V) oxide ≥99%

Supplier: Ambeed
Warning

A608843-100G A608843-5G A608843-25G
77602-544EA 121.98 USD
77602-544 77602-546 77873-000
Tantalum(V) oxide ≥99%
Tantalum(V) oxide

Precaución: for Research Use Only

Formula: O₅Ta₂
MDL Number: MFCD00011254
CAS Number: 1314-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR