5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95+%

Supplier: Ambeed
Warning

A608831-1G A608831-100MG A608831-250MG
77673-650EA 210 USD
77673-650 77884-462 77884-463
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95+%
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MDL Number: MFCD10698508
CAS Number: 1116093-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR