2-(Benzylamino)ethyl hydrogen sulfate ≥95%

Supplier: Ambeed
Danger

A608554-1G A608554-25G
77886-459EA 9 USD
77886-459 77886-460
2-(Benzylamino)ethyl hydrogen sulfate ≥95%
2-(Benzylamino)ethyl hydrogen sulfate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₄S
MW: 231.27 g/mol
MDL Number: MFCD23135361
CAS Number: 2133-07-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR