2-Chloro-7-methoxyquinazoline ≥95%

Supplier: Ambeed
Warning

A607560-100MG A607560-250MG A607560-5G
77893-227EA 47 USD
77893-227 77893-228 77893-229
2-Chloro-7-methoxyquinazoline ≥95%
2-Chloro-7-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD09907933
CAS Number: 953039-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR